Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDIT4
Entrez Gene ID
54541
Gene Name
DNA damage inducible transcript 4
Gene Aliases
Dig2, REDD-1, REDD1
Chromosome Location
Chr.10: 72273919 - 72276039 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019058.2 NP_061931.1 2 - 3 411 68
GenBank mRNA AF335324.1 - 2 - 3 427 68
AK000507.1 - 2 - 3 406 68
AL136668.1 - 2 - 3 410 68
AY090097.1 - 2 - 3 327 68
BC000708.1 - 2 - 3 317 68
BC007714.1 - 2 - 3 399 68
BC015236.1 - 2 - 3 385 68

View Full Product Details