The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032801.4 | NP_116190.3 | 3 - 4 | 412 | 135 |
GenBank mRNA | AF356518.1 | - | 3 - 4 | 253 | 135 |
AF448478.1 | - | 3 - 4 | 287 | 135 | |
AJ344431.1 | - | 3 - 4 | 268 | 135 | |
AJ416101.1 | - | 3 - 4 | 412 | 135 | |
AK074769.1 | - | 3 - 4 | 295 | 135 | |
AK075309.1 | - | 3 - 4 | 266 | 135 | |
AK225666.1 | - | 3 - 4 | 266 | 135 | |
AY358335.1 | - | 3 - 4 | 303 | 135 | |
BC010690.1 | - | 3 - 4 | 251 | 135 | |
BC012147.1 | - | 3 - 4 | 253 | 135 | |
BC057284.1 | - | 3 - 4 | 266 | 135 |