The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031849.2 | NP_001027019.1 | 8 - 9 | 1484 | 90 |
XM_006713701.2 | XP_006713764.1 | 9 - 10 | 1557 | 90 | |
XM_011512991.2 | XP_011511293.1 | 8 - 9 | 1473 | 90 | |
GenBank mRNA | AK129893.1 | - | 8 - 9 | 1183 | 90 |
AK300663.1 | - | 9 - 10 | 1305 | 90 | |
BC039724.1 | - | 8 - 9 | 1419 | 90 | |
CR749615.1 | - | 8 - 9 | 1386 | 90 |