The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324400.1 | NP_001311329.1 | 15 - 16 | 1872 | 155 |
NM_022444.3 | NP_071889.2 | 12 - 13 | 1371 | 155 | |
XM_011516515.2 | XP_011514817.1 | 13 - 14 | 1543 | 155 | |
XM_011516516.2 | XP_011514818.1 | 12 - 13 | 1441 | 155 | |
XM_011516517.2 | XP_011514819.1 | 14 - 15 | 1662 | 155 | |
XM_017012554.1 | XP_016868043.1 | 12 - 13 | 1429 | 155 | |
GenBank mRNA | AF260824.1 | - | 12 - 13 | 1371 | 155 |
AK314537.1 | - | 12 - 13 | 1369 | 155 | |
BC111775.1 | - | 15 - 16 | 1872 | 155 |