The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324400.1 | NP_001311329.1 | 5 - 6 | 730 | 117 |
NM_022444.3 | NP_071889.2 | 4 - 5 | 574 | 117 | |
XM_011516515.2 | XP_011514817.1 | 4 - 5 | 578 | 117 | |
XM_011516516.2 | XP_011514818.1 | 4 - 5 | 578 | 117 | |
XM_011516517.2 | XP_011514819.1 | 5 - 6 | 697 | 117 | |
XM_011516518.2 | XP_011514820.1 | 4 - 5 | 578 | 117 | |
XM_011516519.2 | XP_011514821.1 | 4 - 5 | 578 | 117 | |
XM_017012554.1 | XP_016868043.1 | 3 - 4 | 464 | 117 | |
XM_017012555.1 | XP_016868044.1 | 4 - 5 | 578 | 117 | |
GenBank mRNA | AF260824.1 | - | 4 - 5 | 574 | 117 |
AK026413.1 | - | 4 - 5 | 572 | 117 | |
AK314537.1 | - | 4 - 5 | 572 | 117 | |
BC111775.1 | - | 5 - 6 | 730 | 117 |