Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAAM2
Entrez Gene ID
23500
Gene Name
dishevelled associated activator of morphogenesis 2
Gene Aliases
dJ90A20A.1
Chromosome Location
Chr.6: 39792366 - 39904877 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201427.1 NP_001188356.1 19 - 20 2480 119
NM_015345.3 NP_056160.2 19 - 20 2544 119
XM_006715039.3 XP_006715102.3 20 - 21 3152 119
XM_006715040.3 XP_006715103.3 19 - 20 3124 119
XM_006715042.2 XP_006715105.1 20 - 21 5147 119
XM_006715043.2 XP_006715106.1 20 - 21 2524 119
XM_006715045.3 XP_006715108.1 20 - 21 3739 119
XM_006715046.3 XP_006715109.1 20 - 21 2917 119
XM_017010630.1 XP_016866119.1 20 - 21 3763 119
GenBank mRNA AB002379.2 - 19 - 20 2480 119
AL833083.2 - 19 - 20 2524 119
BX647839.1 - 3 - 4 3062 119

View Full Product Details