Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DAAM2
Entrez Gene ID
23500
Gene Name
dishevelled associated activator of morphogenesis 2
Gene Aliases
dJ90A20A.1
Chromosome Location
Chr.6: 39792366 - 39904877 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201427.1 NP_001188356.1 20 - 21 2645 76
NM_015345.3 NP_056160.2 20 - 21 2709 76
XM_006715039.3 XP_006715102.3 21 - 22 3317 76
XM_006715040.3 XP_006715103.3 20 - 21 3289 76
XM_006715042.2 XP_006715105.1 21 - 22 5312 76
XM_006715043.2 XP_006715106.1 21 - 22 2689 76
XM_006715045.3 XP_006715108.1 21 - 22 3904 76
XM_006715046.3 XP_006715109.1 21 - 22 3082 76
XM_017010630.1 XP_016866119.1 21 - 22 3928 76
GenBank mRNA AB002379.2 - 20 - 21 2645 76
AL833083.2 - 20 - 21 2689 76
BX647839.1 - 4 - 5 3227 76

View Full Product Details