The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032811.2 | NP_116200.2 | 1 - 2 | 387 | 101 |
NR_016021.1 | 1 - 2 | 387 | 101 | ||
GenBank mRNA | AK027527.1 | - | 1 - 2 | 358 | 101 |
AK074140.1 | - | 1 - 2 | 110 | 101 | |
AK222936.1 | - | 1 - 2 | 363 | 101 | |
BC018452.1 | - | 1 - 2 | 355 | 101 | |
BC041043.1 | - | 1 - 2 | 372 | 101 | |
BC064392.1 | - | 1 - 2 | 336 | 101 | |
BC073160.1 | - | 1 - 2 | 316 | 101 | |
BC107428.1 | - | 1 - 2 | 377 | 101 | |
BC109269.2 | - | 1 - 2 | 239 | 101 | |
DA292015.1 | - | 1 - 2 | 387 | 101 | |
DQ144542.1 | - | 1 - 2 | 149 | 101 |