The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202423.1 | NP_001189352.1 | 12 - 13 | 1638 | 109 |
NM_030934.4 | NP_112196.3 | 12 - 13 | 2030 | 109 | |
XM_011510028.2 | XP_011508330.1 | 12 - 13 | 3467 | 109 | |
GenBank mRNA | AF163261.1 | - | 4 - 5 | 493 | 109 |
AF288399.1 | - | 12 - 13 | 1952 | 109 | |
AK074936.1 | - | 12 - 13 | 1979 | 109 | |
AK074993.1 | - | 12 - 13 | 2002 | 109 | |
AK098359.1 | - | 12 - 13 | 1997 | 109 | |
AK131460.1 | - | 12 - 13 | 2737 | 109 | |
AK226103.1 | - | 12 - 13 | 1745 | 109 | |
AK302166.1 | - | 13 - 14 | 1562 | 109 | |
AK312801.1 | - | 12 - 13 | 1999 | 109 | |
BC035430.1 | - | 12 - 13 | 1982 | 109 | |
BC045535.1 | - | 12 - 13 | 1979 | 109 | |
BC053961.1 | - | 12 - 13 | 1979 | 109 |