The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000312.3 | NP_000303.1 | 1 - 2 | 75 | 114 |
XM_017004505.1 | XP_016859994.1 | 1 - 2 | 274 | 114 | |
GenBank mRNA | X02750.1 | - | 1 - 2 | 78 | 114 |