Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TULP2
Entrez Gene ID
7288
Gene Name
tubby like protein 2
Gene Aliases
CT65, TUBL2
Chromosome Location
Chr.19: 48880817 - 48900177 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003323.2 NP_003314.2 12 - 13 1595 71
GenBank mRNA BC026070.2 - 12 - 13 1589 71
U82469.1 - 12 - 13 1537 71

View Full Product Details