Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TULP2
Entrez Gene ID
7288
Gene Name
tubby like protein 2
Gene Aliases
CT65, TUBL2
Chromosome Location
Chr.19: 48880817 - 48900177 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003323.2 NP_003314.2 4 - 5 359 74
XM_006723347.3 XP_006723410.1 4 - 5 359 74
XM_006723348.3 XP_006723411.1 4 - 5 303 74
XM_011527252.2 XP_011525554.1 5 - 6 463 74
XM_011527254.2 XP_011525556.1 4 - 5 359 74
XM_011527255.2 XP_011525557.1 3 - 4 307 74
XM_011527256.2 XP_011525558.1 2 - 3 274 74
GenBank mRNA BC026070.2 - 4 - 5 353 74
U82469.1 - 4 - 5 301 74

View Full Product Details