The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003323.2 | NP_003314.2 | 5 - 6 | 497 | 139 |
XM_006723347.3 | XP_006723410.1 | 5 - 6 | 497 | 139 | |
XM_006723348.3 | XP_006723411.1 | 5 - 6 | 441 | 139 | |
XM_011527252.2 | XP_011525554.1 | 6 - 7 | 601 | 139 | |
XM_011527253.2 | XP_011525555.1 | 5 - 6 | 488 | 139 | |
XM_011527254.2 | XP_011525556.1 | 5 - 6 | 497 | 139 | |
XM_011527255.2 | XP_011525557.1 | 4 - 5 | 445 | 139 | |
XM_011527256.2 | XP_011525558.1 | 3 - 4 | 412 | 139 | |
GenBank mRNA | BC026070.2 | - | 5 - 6 | 491 | 139 |
U82469.1 | - | 5 - 6 | 439 | 139 |