The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206794.1 | NP_001193723.1 | 13 - 14 | 1449 | 151 |
NM_016374.5 | NP_057458.4 | 13 - 14 | 1570 | 151 | |
NM_031371.3 | NP_112739.2 | 13 - 14 | 1570 | 151 | |
NR_135074.1 | 13 - 14 | 1572 | 151 | ||
XM_006711781.2 | XP_006711844.1 | 13 - 14 | 1593 | 151 | |
XM_011544212.2 | XP_011542514.1 | 13 - 14 | 1409 | 151 | |
XM_017001468.1 | XP_016856957.1 | 12 - 13 | 1231 | 151 | |
XM_017001469.1 | XP_016856958.1 | 12 - 13 | 1232 | 151 | |
XM_017001470.1 | XP_016856959.1 | 12 - 13 | 1230 | 151 | |
XM_017001471.1 | XP_016856960.1 | 12 - 13 | 1234 | 151 | |
XM_017001472.1 | XP_016856961.1 | 3 - 4 | 243 | 151 | |
GenBank mRNA | AB210032.1 | - | 13 - 14 | 1449 | 151 |
AY220790.1 | - | 11 - 12 | 1072 | 151 | |
BC017800.1 | - | 3 - 4 | 298 | 151 | |
BC048959.1 | - | 13 - 14 | 1531 | 151 | |
BC062536.1 | - | 13 - 14 | 1520 | 151 | |
BC104632.1 | - | 11 - 12 | 1079 | 151 | |
BC130418.1 | - | 13 - 14 | 1487 | 151 | |
BX648820.1 | - | 13 - 14 | 1522 | 151 |