The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206794.1 | NP_001193723.1 | 5 - 6 | 645 | 91 |
NM_016374.5 | NP_057458.4 | 5 - 6 | 766 | 91 | |
NM_031371.3 | NP_112739.2 | 5 - 6 | 766 | 91 | |
NR_135074.1 | 5 - 6 | 768 | 91 | ||
XM_006711781.2 | XP_006711844.1 | 5 - 6 | 789 | 91 | |
XM_011544212.2 | XP_011542514.1 | 5 - 6 | 605 | 91 | |
XM_017001468.1 | XP_016856957.1 | 4 - 5 | 427 | 91 | |
XM_017001469.1 | XP_016856958.1 | 4 - 5 | 428 | 91 | |
XM_017001470.1 | XP_016856959.1 | 4 - 5 | 426 | 91 | |
XM_017001471.1 | XP_016856960.1 | 4 - 5 | 430 | 91 | |
GenBank mRNA | AB210032.1 | - | 5 - 6 | 645 | 91 |
AF208045.2 | - | 4 - 5 | 312 | 91 | |
AF214114.2 | - | 5 - 6 | 732 | 91 | |
AI356277.1 | - | 5 - 6 | 487 | 91 | |
AK057552.1 | - | 5 - 6 | 766 | 91 | |
BC048959.1 | - | 5 - 6 | 727 | 91 | |
BC062536.1 | - | 5 - 6 | 716 | 91 | |
BC104632.1 | - | 3 - 4 | 275 | 91 | |
BC130418.1 | - | 5 - 6 | 683 | 91 | |
BX648820.1 | - | 5 - 6 | 718 | 91 |