The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018646.5 | NP_061116.5 | 2 - 3 | 460 | 100 |
GenBank mRNA | AF365927.1 | - | 2 - 3 | 447 | 100 |
AJ243500.2 | - | 2 - 3 | 431 | 100 | |
AJ243501.2 | - | 2 - 3 | 431 | 100 | |
AJ487964.2 | - | 3 - 4 | 400 | 100 | |
AK291707.1 | - | 2 - 3 | 461 | 100 | |
KF534785.1 | - | 2 - 3 | 460 | 100 |