The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032998.1 | NP_001028170.1 | 1 - 2 | 106 | 81 |
NM_003937.2 | NP_003928.1 | 1 - 2 | 106 | 81 | |
XM_017005217.1 | XP_016860706.1 | 1 - 2 | 68 | 81 | |
GenBank mRNA | AK223426.1 | - | 1 - 2 | 104 | 81 |
AK290900.1 | - | 1 - 2 | 52 | 81 | |
AK292095.1 | - | 1 - 2 | 70 | 81 | |
AL548658.3 | - | 1 - 2 | 234 | 81 | |
BC000879.1 | - | 1 - 2 | 75 | 81 | |
CB151988.1 | - | 1 - 2 | 106 | 81 | |
U57721.1 | - | 1 - 2 | 82 | 81 |