The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195683.1 | NP_001182612.1 | 16 - 17 | 2944 | 95 |
NM_001195684.1 | NP_001182613.1 | 17 - 18 | 2780 | 95 | |
NM_003243.4 | NP_003234.2 | 16 - 17 | 2947 | 95 | |
NR_036634.1 | 17 - 18 | 3044 | 95 | ||
XM_006710867.2 | XP_006710930.1 | 16 - 17 | 2666 | 95 | |
XM_006710868.2 | XP_006710931.1 | 16 - 17 | 2632 | 95 | |
GenBank mRNA | AJ251961.1 | - | 15 - 16 | 2494 | 95 |
AK225799.1 | - | 16 - 17 | 2722 | 95 | |
AK291345.1 | - | 16 - 17 | 2588 | 95 | |
BC099914.1 | - | 19 - 20 | 2937 | 95 | |
BC126116.1 | - | 16 - 17 | 2600 | 95 | |
BC136295.1 | - | 16 - 17 | 2600 | 95 | |
BC143869.1 | - | 17 - 18 | 2697 | 95 | |
HG764816.1 | - | 17 - 18 | 2780 | 95 | |
L07594.1 | - | 17 - 18 | 2774 | 95 |