The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004997.2 | NP_004988.2 | 8 - 9 | 1295 | 67 |
GenBank mRNA | BC044226.1 | - | 8 - 9 | 1295 | 67 |