Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB17
Entrez Gene ID
7709
Gene Name
zinc finger and BTB domain containing 17
Gene Aliases
MIZ-1, ZNF151, ZNF60, pHZ-67
Chromosome Location
Chr.1: 15941869 - 15976132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242884.1 NP_001229813.1 6 - 7 975 58
NM_001287603.1 NP_001274532.1 7 - 8 1160 58
NM_001287604.1 NP_001274533.1 7 - 8 1156 58
NM_001324137.1 NP_001311066.1 6 - 7 953 58
NM_001324138.1 NP_001311067.1 6 - 7 971 58
NM_003443.2 NP_003434.2 7 - 8 1160 58
XM_005245986.1 XP_005246043.1 6 - 7 971 58
XM_005245987.1 XP_005246044.1 7 - 8 1101 58
XM_011542085.2 XP_011540387.1 6 - 7 946 58
XM_011542086.1 XP_011540388.1 7 - 8 1127 58
XM_011542087.2 XP_011540389.1 6 - 7 968 58
XM_011542088.1 XP_011540390.1 7 - 8 1127 58
XM_017002242.1 XP_016857731.1 5 - 6 941 58
GenBank mRNA AK223618.1 - 7 - 8 1137 58
AK226168.1 - 7 - 8 1100 58
AK299820.1 - 6 - 7 928 58
AK301896.1 - 6 - 7 922 58
AK302610.1 - 6 - 7 900 58
AK303823.1 - 6 - 7 866 58
AK315201.1 - 7 - 8 1107 58
BC126163.1 - 6 - 7 1023 58
BC143965.1 - 6 - 7 1023 58
Y09723.1 - 7 - 8 1047 58

View Full Product Details