The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242884.1 | NP_001229813.1 | 7 - 8 | 1123 | 82 |
NM_001287603.1 | NP_001274532.1 | 8 - 9 | 1308 | 82 | |
NM_001287604.1 | NP_001274533.1 | 8 - 9 | 1304 | 82 | |
NM_001324137.1 | NP_001311066.1 | 7 - 8 | 1101 | 82 | |
NM_001324138.1 | NP_001311067.1 | 7 - 8 | 1119 | 82 | |
NM_003443.2 | NP_003434.2 | 8 - 9 | 1308 | 82 | |
XM_005245986.1 | XP_005246043.1 | 7 - 8 | 1119 | 82 | |
XM_005245987.1 | XP_005246044.1 | 8 - 9 | 1249 | 82 | |
XM_011542085.2 | XP_011540387.1 | 7 - 8 | 1094 | 82 | |
XM_011542086.1 | XP_011540388.1 | 8 - 9 | 1275 | 82 | |
XM_011542087.2 | XP_011540389.1 | 7 - 8 | 1116 | 82 | |
XM_011542088.1 | XP_011540390.1 | 8 - 9 | 1275 | 82 | |
XM_017002242.1 | XP_016857731.1 | 6 - 7 | 1089 | 82 | |
GenBank mRNA | AK223618.1 | - | 8 - 9 | 1285 | 82 |
AK226168.1 | - | 8 - 9 | 1248 | 82 | |
AK299820.1 | - | 7 - 8 | 1076 | 82 | |
AK301896.1 | - | 7 - 8 | 1070 | 82 | |
AK303823.1 | - | 7 - 8 | 1014 | 82 | |
AK315201.1 | - | 8 - 9 | 1255 | 82 | |
BC126163.1 | - | 7 - 8 | 1171 | 82 | |
BC143965.1 | - | 7 - 8 | 1171 | 82 | |
Y09723.1 | - | 8 - 9 | 1195 | 82 |