Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPP7
Entrez Gene ID
29952
Gene Name
dipeptidyl peptidase 7
Gene Aliases
DPP2, DPPII, QPP
Chromosome Location
Chr.9: 137110540 - 137117980 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013379.2 NP_037511.2 4 - 5 509 97
XM_011518599.2 XP_011516901.1 3 - 4 560 97
XM_011518600.1 XP_011516902.1 4 - 5 470 97
XM_017014651.1 XP_016870140.1 3 - 4 560 97
XM_017014652.1 XP_016870141.1 4 - 5 520 97
GenBank mRNA AB209787.1 - 4 - 5 476 97
AF154502.1 - 4 - 5 479 97
AK292112.1 - 4 - 5 486 97
AK293700.1 - 3 - 4 552 97
AK298288.1 - 3 - 4 342 97
AL533195.3 - 4 - 5 490 97
BC011907.2 - 4 - 5 484 97
BC016961.1 - 4 - 5 484 97
BX400716.2 - 4 - 5 507 97

View Full Product Details