Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCOR3
Entrez Gene ID
55758
Gene Name
REST corepressor 3
Gene Aliases
-
Chromosome Location
Chr.1: 211259347 - 211316385 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136223.1 NP_001129695.1 4 - 5 546 84
NM_001136224.2 NP_001129696.2 4 - 5 546 84
NM_001136225.1 NP_001129697.1 4 - 5 546 84
NM_018254.3 NP_060724.1 3 - 4 346 84
XM_005273191.4 XP_005273248.1 4 - 5 530 84
XM_006711442.2 XP_006711505.1 4 - 5 530 84
XM_011509748.2 XP_011508050.1 4 - 5 530 84
XM_011509749.2 XP_011508051.1 4 - 5 530 84
XM_017001758.1 XP_016857247.1 5 - 6 664 84
XM_017001759.1 XP_016857248.1 5 - 6 639 84
XM_017001760.1 XP_016857249.1 5 - 6 664 84
XM_017001761.1 XP_016857250.1 4 - 5 546 84
XM_017001762.1 XP_016857251.1 5 - 6 664 84
XM_017001763.1 XP_016857252.1 5 - 6 664 84
XM_017001764.1 XP_016857253.1 4 - 5 546 84
XM_017001765.1 XP_016857254.1 5 - 6 664 84
XM_017001766.1 XP_016857255.1 5 - 6 874 84
XM_017001767.1 XP_016857256.1 5 - 6 664 84
GenBank mRNA AB037764.1 - 3 - 4 306 84
AK001738.1 - 3 - 4 318 84
AK131312.1 - 4 - 5 524 84
AK300942.1 - 5 - 6 612 84
AK302664.1 - 4 - 5 479 84
BC031608.1 - 4 - 5 398 84
DA478986.1 - 3 - 4 346 84

View Full Product Details