Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCOR3
Entrez Gene ID
55758
Gene Name
REST corepressor 3
Gene Aliases
-
Chromosome Location
Chr.1: 211259347 - 211316385 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136223.1 NP_001129695.1 6 - 7 837 152
NM_001136224.2 NP_001129696.2 6 - 7 837 152
NM_001136225.1 NP_001129697.1 6 - 7 837 152
NM_018254.3 NP_060724.1 5 - 6 637 152
XM_005273191.4 XP_005273248.1 6 - 7 821 152
XM_006711442.2 XP_006711505.1 6 - 7 821 152
XM_011509748.2 XP_011508050.1 6 - 7 821 152
XM_011509749.2 XP_011508051.1 6 - 7 821 152
XM_017001758.1 XP_016857247.1 7 - 8 955 152
XM_017001759.1 XP_016857248.1 7 - 8 930 152
XM_017001760.1 XP_016857249.1 7 - 8 955 152
XM_017001761.1 XP_016857250.1 6 - 7 837 152
XM_017001762.1 XP_016857251.1 7 - 8 955 152
XM_017001763.1 XP_016857252.1 7 - 8 955 152
XM_017001764.1 XP_016857253.1 6 - 7 837 152
XM_017001765.1 XP_016857254.1 7 - 8 955 152
XM_017001766.1 XP_016857255.1 7 - 8 1165 152
XM_017001767.1 XP_016857256.1 7 - 8 955 152
GenBank mRNA AB037764.1 - 5 - 6 597 152
AK001738.1 - 5 - 6 609 152
AK131312.1 - 6 - 7 815 152
AK300942.1 - 7 - 8 903 152
AK302664.1 - 6 - 7 770 152
BC031608.1 - 6 - 7 689 152

View Full Product Details