The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182559.2 | NP_872365.1 | 3 - 4 | 695 | 137 |
GenBank mRNA | BC048112.1 | - | 3 - 4 | 680 | 137 |