Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MECOM
- Entrez Gene ID
-
2122
- Gene Name
- MDS1 and EVI1 complex locus
- Gene Aliases
-
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
- Chromosome Location
-
Chr.3: 169083499 - 169663781 on Build GRCh38
View Full Product Details