The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105077.3 | NP_001098547.3 | 10 - 11 | 2373 | 72 |
NM_001105078.3 | NP_001098548.2 | 9 - 10 | 2364 | 72 | |
NM_001205194.1 | NP_001192123.1 | 9 - 10 | 2668 | 72 | |
NM_004991.3 | NP_004982.2 | 10 - 11 | 3006 | 72 | |
NM_005241.3 | NP_005232.2 | 9 - 10 | 2308 | 72 | |
XM_005247213.3 | XP_005247270.1 | 10 - 11 | 3015 | 72 | |
XM_005247219.2 | XP_005247276.1 | 9 - 10 | 2980 | 72 | |
XM_005247220.2 | XP_005247277.1 | 9 - 10 | 2321 | 72 | |
XM_005247221.2 | XP_005247278.1 | 9 - 10 | 2304 | 72 | |
XM_005247223.2 | XP_005247280.1 | 9 - 10 | 2977 | 72 | |
XM_005247224.3 | XP_005247281.1 | 10 - 11 | 2043 | 72 | |
XM_005247225.3 | XP_005247282.1 | 10 - 11 | 2039 | 72 | |
XM_011512546.2 | XP_011510848.1 | 9 - 10 | 2768 | 72 | |
XM_017005874.1 | XP_016861363.1 | 9 - 10 | 3683 | 72 | |
GenBank mRNA | AK025934.1 | - | 3 - 4 | 212 | 72 |
AK292865.1 | - | 9 - 10 | 2291 | 72 | |
AK304098.1 | - | 10 - 11 | 2952 | 72 | |
BC130520.1 | - | 10 - 11 | 2347 | 72 | |
BC143951.1 | - | 10 - 11 | 2347 | 72 | |
GQ352634.1 | - | 9 - 10 | 2572 | 72 | |
X54989.1 | - | 9 - 10 | 2306 | 72 |