Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTSL2
Entrez Gene ID
9719
Gene Name
ADAMTS like 2
Gene Aliases
GPHYSD1
Chromosome Location
Chr.9: 133532164 - 133575519 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145320.1 NP_001138792.1 12 - 13 2301 99
NM_014694.3 NP_055509.2 12 - 13 1959 99
XM_005272237.3 XP_005272294.1 12 - 13 2156 99
XM_005272238.3 XP_005272295.1 11 - 12 1798 99
XM_005272239.2 XP_005272296.1 12 - 13 1834 99
XM_011519241.2 XP_011517543.2 11 - 12 1977 99
XM_011519242.2 XP_011517544.1 11 - 12 1833 99
GenBank mRNA AB011177.1 - 12 - 13 2301 99
AK293498.1 - 2 - 3 329 99
BC050544.1 - 12 - 13 1959 99

View Full Product Details