Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TKT
Entrez Gene ID
7086
Gene Name
transketolase
Gene Aliases
HEL-S-48, HEL107, TK, TKT1
Chromosome Location
Chr.3: 53224707 - 53256114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001064.3 NP_001055.1 11 - 12 1651 88
NM_001135055.2 NP_001128527.1 11 - 12 1651 88
NM_001258028.1 NP_001244957.1 12 - 13 1675 88
NR_047580.1 11 - 12 1541 88
XM_011534054.1 XP_011532356.1 12 - 13 1613 88
XM_011534055.1 XP_011532357.1 12 - 13 1700 88
GenBank mRNA AK057092.1 - 5 - 6 1480 88
AK092084.1 - 6 - 7 2514 88
AK093667.1 - 10 - 11 1310 88
AK223614.1 - 11 - 12 1559 88
AK289454.1 - 11 - 12 1527 88
AK293438.1 - 12 - 13 1583 88
AK301231.1 - 11 - 12 1449 88
AK303191.1 - 12 - 13 1518 88
BC002433.2 - 12 - 13 1440 88
BC008615.1 - 6 - 7 795 88
BC009970.2 - 11 - 12 1549 88
BC024026.2 - 9 - 10 1220 88
BQ430230.1 - 2 - 3 199 88
BX647248.1 - 11 - 12 1537 88
BX649193.1 - 5 - 6 4788 88
CA421169.1 - 3 - 4 88
EU794667.1 - 11 - 12 1549 88
GQ472197.1 - 6 - 7 795 88
KU178361.1 - 11 - 12 1479 88
KU178363.1 - 4 - 5 365 88
L12711.1 - 11 - 12 1577 88
U55017.1 - 11 - 12 1479 88
X67688.1 - 12 - 13 1549 88

View Full Product Details