Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCTP1
Entrez Gene ID
79772
Gene Name
multiple C2 and transmembrane domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 94703690 - 95284575 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002796.3 NP_001002796.1 9 - 10 1243 126
NM_001297777.1 NP_001284706.1 7 - 8 1105 126
NM_024717.5 NP_078993.4 9 - 10 1522 126
XM_005272082.4 XP_005272139.1 10 - 11 1649 126
XM_005272090.4 XP_005272147.1 7 - 8 884 126
XM_006714692.3 XP_006714755.1 7 - 8 1472 126
XM_011543649.2 XP_011541951.1 10 - 11 1723 126
XM_011543650.2 XP_011541952.1 8 - 9 700 126
XM_017009855.1 XP_016865344.1 8 - 9 923 126
XM_017009856.1 XP_016865345.1 9 - 10 1022 126
XM_017009857.1 XP_016865346.1 8 - 9 769 126
XM_017009858.1 XP_016865347.1 7 - 8 845 126
XM_017009859.1 XP_016865348.1 9 - 10 847 126
XM_017009860.1 XP_016865349.1 8 - 9 944 126
XM_017009861.1 XP_016865350.1 7 - 8 1219 126
XM_017009862.1 XP_016865351.1 6 - 7 1217 126
XM_017009863.1 XP_016865352.1 6 - 7 845 126
XM_017009864.1 XP_016865353.1 8 - 9 1585 126
XM_017009865.1 XP_016865354.1 7 - 8 1165 126
GenBank mRNA AK057694.1 - 7 - 8 884 126
AK058012.1 - 8 - 9 789 126
AK091330.1 - 2 - 3 246 126
AY656715.1 - 9 - 10 1522 126
AY656716.1 - 9 - 10 859 126
BC030005.2 - 9 - 10 1004 126

View Full Product Details