The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000262.2 | NP_000253.1 | 1 - 2 | 562 | 60 |
XM_005261615.4 | XP_005261672.1 | 2 - 3 | 201 | 60 | |
XM_005261616.4 | XP_005261673.1 | 2 - 3 | 183 | 60 | |
GenBank mRNA | BC000095.2 | - | 1 - 2 | 98 | 60 |
CB988165.1 | - | 1 - 2 | 592 | 60 | |
CR456527.1 | - | 1 - 2 | 71 | 60 | |
M29276.1 | - | 1 - 2 | 360 | 60 | |
M38083.1 | - | 1 - 2 | 89 | 60 | |
M62783.1 | - | 1 - 2 | 486 | 60 |