The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005095.2 | NP_005086.2 | 22 - 23 | 3518 | 70 |
XM_005271328.4 | XP_005271385.1 | 22 - 23 | 3797 | 70 | |
XM_005271331.2 | XP_005271388.1 | 19 - 20 | 2763 | 70 | |
XM_011542421.2 | XP_011540723.1 | 22 - 23 | 3444 | 70 | |
XM_011542423.2 | XP_011540725.1 | 23 - 24 | 3849 | 70 | |
XM_011542425.2 | XP_011540727.1 | 20 - 21 | 2803 | 70 | |
XM_017002803.1 | XP_016858292.1 | 23 - 24 | 3571 | 70 | |
XM_017002804.1 | XP_016858293.1 | 23 - 24 | 3511 | 70 | |
XM_017002805.1 | XP_016858294.1 | 18 - 19 | 3009 | 70 | |
GenBank mRNA | AB007885.1 | - | 18 - 19 | 2638 | 70 |
AK309705.1 | - | 21 - 22 | 3178 | 70 | |
BC012093.1 | - | 20 - 21 | 3097 | 70 | |
BC127113.1 | - | 23 - 24 | 3524 | 70 | |
BC127114.1 | - | 19 - 20 | 2707 | 70 | |
BX647376.1 | - | 18 - 19 | 2749 | 70 | |
BX648067.1 | - | 13 - 14 | 1812 | 70 |