The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005095.2 | NP_005086.2 | 26 - 27 | 4026 | 132 |
XM_005271328.4 | XP_005271385.1 | 26 - 27 | 4305 | 132 | |
XM_005271331.2 | XP_005271388.1 | 23 - 24 | 3271 | 132 | |
XM_011542421.2 | XP_011540723.1 | 26 - 27 | 3952 | 132 | |
XM_011542423.2 | XP_011540725.1 | 27 - 28 | 4357 | 132 | |
XM_011542425.2 | XP_011540727.1 | 24 - 25 | 3311 | 132 | |
XM_017002803.1 | XP_016858292.1 | 27 - 28 | 4079 | 132 | |
XM_017002804.1 | XP_016858293.1 | 27 - 28 | 4019 | 132 | |
XM_017002805.1 | XP_016858294.1 | 22 - 23 | 3517 | 132 | |
GenBank mRNA | AB007885.1 | - | 22 - 23 | 3146 | 132 |
AK309705.1 | - | 25 - 26 | 3686 | 132 | |
BC012093.1 | - | 24 - 25 | 3605 | 132 | |
BC127113.1 | - | 27 - 28 | 4032 | 132 | |
BC127114.1 | - | 23 - 24 | 3215 | 132 | |
BX647376.1 | - | 22 - 23 | 3257 | 132 | |
BX648067.1 | - | 17 - 18 | 2320 | 132 | |
CR936741.1 | - | 25 - 26 | 3890 | 132 |