The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005095.2 | NP_005086.2 | 2 - 3 | 169 | 125 |
XM_017002804.1 | XP_016858293.1 | 2 - 3 | 86 | 125 | |
GenBank mRNA | AK308320.1 | - | 2 - 3 | 169 | 125 |
BG499626.1 | - | 2 - 3 | 170 | 125 |