The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003757.3 | NP_003748.1 | 10 - 11 | 1335 | 63 |
XM_017002671.1 | XP_016858160.1 | 7 - 8 | 608 | 63 | |
GenBank mRNA | AF116697.1 | - | 2 - 3 | 99 | 63 |
AK130633.1 | - | 9 - 10 | 833 | 63 | |
AK222483.1 | - | 9 - 10 | 837 | 63 | |
AK225024.1 | - | 9 - 10 | 823 | 63 | |
AK289356.1 | - | 9 - 10 | 848 | 63 | |
AK289882.1 | - | 10 - 11 | 1335 | 63 | |
BC000413.2 | - | 9 - 10 | 816 | 63 | |
BC003140.1 | - | 8 - 9 | 820 | 63 | |
BC018815.2 | - | 9 - 10 | 816 | 63 | |
BT019597.1 | - | 8 - 9 | 802 | 63 | |
U36764.1 | - | 9 - 10 | 839 | 63 | |
U39067.1 | - | 9 - 10 | 819 | 63 |