The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000419.4 | NP_000410.2 | 23 - 24 | 2476 | 127 |
XM_011524749.1 | XP_011523051.1 | 23 - 24 | 2380 | 127 | |
XM_011524750.1 | XP_011523052.1 | 23 - 24 | 2380 | 127 | |
GenBank mRNA | AB209552.1 | - | 11 - 12 | 1143 | 127 |
AF098114.1 | - | 1 - 2 | 93 | 127 | |
AK315335.1 | - | 23 - 24 | 2380 | 127 | |
BC117443.1 | - | 23 - 24 | 2374 | 127 | |
BC126442.1 | - | 23 - 24 | 2374 | 127 | |
J02764.1 | - | 23 - 24 | 2349 | 127 | |
M34480.1 | - | 23 - 24 | 2380 | 127 | |
X06831.1 | - | 12 - 13 | 1175 | 127 |