Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC3
Entrez Gene ID
51304
Gene Name
zinc finger DHHC-type containing 3
Gene Aliases
DHHC-3, GODZ, ZNF373
Chromosome Location
Chr.3: 44915261 - 44976190 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135179.1 NP_001128651.1 1 - 2 245 160
NM_016598.2 NP_057682.1 1 - 2 245 160
XM_005265209.1 XP_005265266.1 1 - 2 245 160
XM_005265212.2 XP_005265269.1 1 - 2 253 160
XM_006713189.1 XP_006713252.1 1 - 2 245 160
XM_006713192.2 XP_006713255.1 1 - 2 245 160
XM_006713193.2 XP_006713256.1 1 - 2 246 160
XM_006713194.3 XP_006713257.1 1 - 2 224 160
XM_011533797.1 XP_011532099.1 1 - 2 245 160
XM_011533800.2 XP_011532102.1 1 - 2 242 160
XM_011533801.2 XP_011532103.1 1 - 2 245 160
XM_017006557.1 XP_016862046.1 1 - 2 245 160
XM_017006560.1 XP_016862049.1 1 - 2 245 160
XM_017006561.1 XP_016862050.1 1 - 2 245 160
XM_017006562.1 XP_016862051.1 1 - 2 245 160
XM_017006563.1 XP_016862052.1 1 - 2 245 160
XM_017006564.1 XP_016862053.1 1 - 2 245 160
XM_017006565.1 XP_016862054.1 1 - 2 230 160
XM_017006569.1 XP_016862058.1 1 - 2 222 160
XM_017006570.1 XP_016862059.1 1 - 2 228 160
XM_017006571.1 XP_016862060.1 1 - 2 237 160
GenBank mRNA AF247703.1 - 1 - 2 185 160
BC015467.1 - 1 - 2 172 160
DB636483.1 - 1 - 2 245 160

View Full Product Details