Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD17
Entrez Gene ID
79734
Gene Name
potassium channel tetramerization domain containing 17
Gene Aliases
-
Chromosome Location
Chr.22: 37051725 - 37063390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282684.1 NP_001269613.1 1 - 2 214 94
NM_001282685.1 NP_001269614.1 1 - 2 214 94
NM_001282686.1 NP_001269615.1 1 - 2 214 94
NM_024681.3 NP_078957.2 1 - 2 214 94
XM_005261741.3 XP_005261798.2 1 - 2 225 94
XM_005261742.2 XP_005261799.2 1 - 2 225 94
XM_005261743.2 XP_005261800.2 1 - 2 225 94
XM_005261744.2 XP_005261801.1 1 - 2 225 94
XM_011530374.2 XP_011528676.1 1 - 2 225 94
XM_011530377.2 XP_011528679.1 1 - 2 225 94
GenBank mRNA AK022304.1 - 1 - 2 190 94
BC025403.1 - 1 - 2 213 94
BC031038.1 - 1 - 2 197 94
BG832241.1 - 1 - 2 195 94
BX463485.2 - 1 - 2 74 94
BX951630.1 - 1 - 2 126 94
DA258908.1 - 1 - 2 207 94
HY029628.1 - 2 - 3 139 94
HY034580.1 - 1 - 2 214 94

View Full Product Details