Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM37
Entrez Gene ID
4591
Gene Name
tripartite motif containing 37
Gene Aliases
MUL, POB1, TEF3
Chromosome Location
Chr.17: 58968068 - 59106921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005207.3 NP_001005207.1 11 - 12 1399 65
NM_001320987.1 NP_001307916.1 10 - 11 1297 65
NM_001320988.1 NP_001307917.1 11 - 12 1399 65
NM_001320989.1 NP_001307918.1 11 - 12 1399 65
NM_001320990.1 NP_001307919.1 12 - 13 1475 65
NM_015294.4 NP_056109.1 11 - 12 1399 65
XM_005257385.1 XP_005257442.1 11 - 12 1231 65
XM_011524831.1 XP_011523133.1 11 - 12 1231 65
XM_011524832.2 XP_011523134.1 11 - 12 1231 65
XM_011524833.1 XP_011523135.1 10 - 11 1180 65
XM_011524834.1 XP_011523136.1 10 - 11 1113 65
XM_011524836.1 XP_011523138.1 11 - 12 1231 65
XM_017024662.1 XP_016880151.1 11 - 12 1231 65
XM_017024663.1 XP_016880152.1 11 - 12 1231 65
XM_017024664.1 XP_016880153.1 10 - 11 1180 65
XM_017024665.1 XP_016880154.1 10 - 11 1113 65
XM_017024666.1 XP_016880155.1 10 - 11 1180 65
XM_017024667.1 XP_016880156.1 10 - 11 1180 65
XM_017024668.1 XP_016880157.1 10 - 11 1113 65
XM_017024669.1 XP_016880158.1 9 - 10 1062 65
XM_017024670.1 XP_016880159.1 10 - 11 1112 65
XM_017024671.1 XP_016880160.1 10 - 11 1143 65
XM_017024672.1 XP_016880161.1 10 - 11 1143 65
XM_017024673.1 XP_016880162.1 10 - 11 935 65
XM_017024674.1 XP_016880163.1 10 - 11 935 65
GenBank mRNA AB020705.1 - 11 - 12 984 65
AF213365.1 - 11 - 12 1203 65
AK025648.1 - 5 - 6 417 65
AK289674.1 - 11 - 12 1214 65
AK292459.1 - 11 - 12 1271 65
AK294850.1 - 10 - 11 1202 65
BC036012.1 - 11 - 12 1235 65
BX537955.1 - 12 - 13 1294 65

View Full Product Details