Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM37
Entrez Gene ID
4591
Gene Name
tripartite motif containing 37
Gene Aliases
MUL, POB1, TEF3
Chromosome Location
Chr.17: 58968068 - 59106921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005207.3 NP_001005207.1 3 - 4 627 86
NM_001320988.1 NP_001307917.1 3 - 4 627 86
NM_001320989.1 NP_001307918.1 3 - 4 627 86
NM_015294.4 NP_056109.1 3 - 4 627 86
XM_005257385.1 XP_005257442.1 3 - 4 459 86
XM_011524831.1 XP_011523133.1 3 - 4 459 86
XM_011524832.2 XP_011523134.1 3 - 4 459 86
XM_011524833.1 XP_011523135.1 3 - 4 459 86
XM_011524836.1 XP_011523138.1 3 - 4 459 86
XM_017024662.1 XP_016880151.1 3 - 4 459 86
XM_017024663.1 XP_016880152.1 3 - 4 459 86
XM_017024664.1 XP_016880153.1 3 - 4 459 86
XM_017024666.1 XP_016880155.1 3 - 4 459 86
XM_017024667.1 XP_016880156.1 3 - 4 459 86
XM_017024671.1 XP_016880160.1 3 - 4 459 86
XM_017024672.1 XP_016880161.1 3 - 4 459 86
XM_017024673.1 XP_016880162.1 2 - 3 146 86
XM_017024674.1 XP_016880163.1 2 - 3 146 86
GenBank mRNA AB020705.1 - 3 - 4 212 86
AF213365.1 - 3 - 4 431 86
AK289674.1 - 3 - 4 442 86
AK292459.1 - 3 - 4 499 86
BC036012.1 - 3 - 4 463 86
BG722700.1 - 3 - 4 579 86

View Full Product Details