Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM37
Entrez Gene ID
4591
Gene Name
tripartite motif containing 37
Gene Aliases
MUL, POB1, TEF3
Chromosome Location
Chr.17: 58968068 - 59106921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005207.3 NP_001005207.1 7 - 8 1079 106
NM_001320987.1 NP_001307916.1 6 - 7 977 106
NM_001320988.1 NP_001307917.1 7 - 8 1079 106
NM_001320989.1 NP_001307918.1 7 - 8 1079 106
NM_001320990.1 NP_001307919.1 8 - 9 1155 106
NM_015294.4 NP_056109.1 7 - 8 1079 106
XM_005257385.1 XP_005257442.1 7 - 8 911 106
XM_011524831.1 XP_011523133.1 7 - 8 911 106
XM_011524832.2 XP_011523134.1 7 - 8 911 106
XM_011524833.1 XP_011523135.1 7 - 8 911 106
XM_011524834.1 XP_011523136.1 6 - 7 793 106
XM_011524836.1 XP_011523138.1 7 - 8 911 106
XM_017024662.1 XP_016880151.1 7 - 8 911 106
XM_017024663.1 XP_016880152.1 7 - 8 911 106
XM_017024664.1 XP_016880153.1 7 - 8 911 106
XM_017024665.1 XP_016880154.1 6 - 7 793 106
XM_017024666.1 XP_016880155.1 7 - 8 911 106
XM_017024667.1 XP_016880156.1 7 - 8 911 106
XM_017024668.1 XP_016880157.1 6 - 7 793 106
XM_017024669.1 XP_016880158.1 6 - 7 793 106
XM_017024670.1 XP_016880159.1 6 - 7 792 106
XM_017024671.1 XP_016880160.1 6 - 7 823 106
XM_017024672.1 XP_016880161.1 6 - 7 823 106
XM_017024673.1 XP_016880162.1 6 - 7 598 106
XM_017024674.1 XP_016880163.1 6 - 7 598 106
GenBank mRNA AB020705.1 - 7 - 8 664 106
AF213365.1 - 7 - 8 883 106
AK025648.1 - 1 - 2 97 106
AK289674.1 - 7 - 8 894 106
AK292459.1 - 7 - 8 951 106
BC036012.1 - 7 - 8 915 106
BX537955.1 - 8 - 9 974 106

View Full Product Details