The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014877.3 | NP_055692.2 | 29 - 30 | 4676 | 82 |
XM_005257888.4 | XP_005257945.1 | 30 - 31 | 4805 | 82 | |
XM_005257889.4 | XP_005257946.1 | 29 - 30 | 4727 | 82 | |
XM_006722214.3 | XP_006722277.1 | 30 - 31 | 4808 | 82 | |
XM_006722215.3 | XP_006722278.1 | 28 - 29 | 4680 | 82 | |
XM_006722216.3 | XP_006722279.1 | 19 - 20 | 3510 | 82 | |
XM_011525544.2 | XP_011523846.1 | 30 - 31 | 4879 | 82 | |
XM_017025477.1 | XP_016880966.1 | 27 - 28 | 4596 | 82 | |
XM_017025478.1 | XP_016880967.1 | 30 - 31 | 4861 | 82 | |
GenBank mRNA | AK310639.1 | - | 2 - 3 | 403 | 82 |
BC094881.1 | - | 27 - 28 | 4702 | 82 | |
BC130582.1 | - | 29 - 30 | 4656 | 82 | |
BC144083.1 | - | 29 - 30 | 4659 | 82 | |
D29677.2 | - | 28 - 29 | 4634 | 82 |