Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRMD3
Entrez Gene ID
257019
Gene Name
FERM domain containing 3
Gene Aliases
4.1O, EPB41L4O, EPB41LO, P410
Chromosome Location
Chr.9: 83242990 - 83560208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244959.1 NP_001231888.1 11 - 12 1198 84
NM_001244960.1 NP_001231889.1 11 - 12 974 84
NM_001244961.1 NP_001231890.1 6 - 7 510 84
NM_174938.5 NP_777598.3 11 - 12 1198 84
XM_017014588.1 XP_016870077.1 11 - 12 888 84
XM_017014589.1 XP_016870078.1 6 - 7 3325 84
XM_017014590.1 XP_016870079.1 10 - 11 1111 84
GenBank mRNA AK094281.1 - 11 - 12 899 84
AK223597.1 - 11 - 12 1125 84
AK297722.1 - 11 - 12 1116 84
AY137774.1 - 11 - 12 982 84
BC037253.1 - 6 - 7 510 84
EF560742.1 - 11 - 12 1049 84

View Full Product Details