The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014229.2 | NP_055044.1 | 11 - 12 | 1544 | 84 |
XM_017007073.1 | XP_016862562.1 | 12 - 13 | 1395 | 84 | |
GenBank mRNA | AK312719.1 | - | 11 - 12 | 1544 | 84 |
S75989.1 | - | 11 - 12 | 1512 | 84 |