The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004256.3 | NP_004247.2 | 3 - 4 | 692 | 72 |
GenBank mRNA | AB010438.1 | - | 3 - 4 | 659 | 72 |
AK315301.1 | - | 3 - 4 | 687 | 72 | |
BC035973.1 | - | 3 - 4 | 687 | 72 |