The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004256.3 | NP_004247.2 | 4 - 5 | 859 | 59 |
XM_017007519.1 | XP_016863008.1 | 3 - 4 | 605 | 59 | |
GenBank mRNA | AB010438.1 | - | 4 - 5 | 826 | 59 |
AK298375.1 | - | 3 - 4 | 605 | 59 | |
AK315301.1 | - | 4 - 5 | 854 | 59 | |
BC035973.1 | - | 4 - 5 | 854 | 59 |