Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC28
Entrez Gene ID
23331
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
TPRBK
Chromosome Location
Chr.22: 27978014 - 28679865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145418.1 NP_001138890.1 11 - 12 3906 76
XM_005261405.2 XP_005261462.1 11 - 12 3906 76
XM_006724171.3 XP_006724234.1 10 - 11 12684 76
XM_011530018.2 XP_011528320.1 11 - 12 4085 76
XM_011530019.2 XP_011528321.1 11 - 12 3906 76
XM_011530020.1 XP_011528322.1 11 - 12 3906 76
XM_011530021.2 XP_011528323.1 11 - 12 3906 76
XM_011530022.1 XP_011528324.1 11 - 12 3906 76
XM_017028673.1 XP_016884162.1 11 - 12 3879 76
GenBank mRNA AB028966.3 - 6 - 7 2699 76
AB665284.1 - 11 - 12 3854 76
BC016465.1 - 3 - 4 450 76

View Full Product Details