The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297777.1 | NP_001284706.1 | 5 - 6 | 894 | 105 |
XM_005272090.4 | XP_005272147.1 | 5 - 6 | 673 | 105 | |
XM_017009861.1 | XP_016865350.1 | 5 - 6 | 1008 | 105 | |
XM_017009864.1 | XP_016865353.1 | 6 - 7 | 1374 | 105 | |
XM_017009865.1 | XP_016865354.1 | 5 - 6 | 954 | 105 | |
GenBank mRNA | AK057694.1 | - | 5 - 6 | 673 | 105 |
AK297325.1 | - | 5 - 6 | 598 | 105 |