Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHC3
Entrez Gene ID
80012
Gene Name
polyhomeotic homolog 3
Gene Aliases
EDR3, HPH3
Chromosome Location
Chr.3: 170087580 - 170181779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308116.1 NP_001295045.1 9 - 10 1992 97
NM_024947.3 NP_079223.3 9 - 10 1977 97
XM_005247789.2 XP_005247846.1 10 - 11 2055 97
XM_005247791.3 XP_005247848.1 10 - 11 2055 97
XM_006713753.1 XP_006713816.1 10 - 11 2024 97
XM_006713754.3 XP_006713817.1 10 - 11 2146 97
XM_006713756.2 XP_006713819.1 8 - 9 1797 97
XM_011513186.2 XP_011511488.1 9 - 10 1956 97
XM_011513187.2 XP_011511489.1 8 - 9 1908 97
XM_011513188.2 XP_011511490.1 9 - 10 1896 97
XM_011513189.2 XP_011511491.1 8 - 9 1848 97
XM_017007235.1 XP_016862724.1 10 - 11 2043 97
XM_017007236.1 XP_016862725.1 10 - 11 2031 97
XM_017007237.1 XP_016862726.1 8 - 9 1848 97
XM_017007238.1 XP_016862727.1 7 - 8 1749 97
XM_017007239.1 XP_016862728.1 7 - 8 1596 97
GenBank mRNA AF380154.1 - 9 - 10 1914 97
AF444193.1 - 8 - 9 1929 97
AJ320486.3 - 9 - 10 1971 97
AK304411.1 - 8 - 9 1818 97
BC131772.1 - 9 - 10 1956 97
EF560717.1 - 9 - 10 1952 97

View Full Product Details