Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COPS7B
Entrez Gene ID
64708
Gene Name
COP9 signalosome subunit 7B
Gene Aliases
CSN7B, SGN7b
Chromosome Location
Chr.2: 231781671 - 231809253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282950.2 NP_001269879.1 1 - 2 158 76
NM_001282952.2 NP_001269881.1 1 - 2 158 76
NM_001308381.1 NP_001295310.1 1 - 2 158 76
NM_022730.3 NP_073567.1 1 - 2 158 76
XM_011511639.1 XP_011509941.1 1 - 2 107 76
GenBank mRNA AK022674.1 - 1 - 2 89 76
AK024273.1 - 1 - 2 107 76
AK222879.1 - 1 - 2 96 76
BC091493.1 - 1 - 2 93 76
BX333333.2 - 1 - 2 96 76

View Full Product Details